missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The NUDT15 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human NUDT15. This antibody recognizes human antigen. The NUDT15 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
| Antigen | NUDT15 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Gene Symbols | FLJ10956; MGC104352; MTH2; RP11-90M2.1 |
| Host Species | Rabbit |
| Immunogen | Fusion Protein |
| Purification Method | Antigen affinity purification |
| Quantity | 150 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?