missing translation for 'onlineSavingsMsg'
Learn More

NUDT15 Rabbit Polyclonal Antibody, Proteintech

Product Code. 30237853 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30237853 20 μL 20µL
30237156 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30237853 Supplier Proteintech Supplier No. 259141AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The NUDT15 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human NUDT15. This antibody recognizes human antigen. The NUDT15 antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Antigen NUDT15
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Gene Symbols FLJ10956; MGC104352; MTH2; RP11-90M2.1
Host Species Rabbit
Immunogen Fusion Protein
Purification Method Antigen affinity purification
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55270
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 19 kDa
Product Type Polyclonal Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.