missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT14 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00€
Specifications
| Antigen | NUDT14 |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
NUDT14 Polyclonal specifically detects NUDT14 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| NUDT14 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| EC 3.6.1.45, Nucleoside diphosphate-linked moiety X motif 14, nudix (nucleoside diphosphate linked moiety X)-type motif 14, Nudix motif 14, UDPG pyrophosphatase, UDP-sugar diphosphatase, UGPPase, UGPPnucleoside diphosphate linked moiety X-type motif 14, uridine diphosphate glucose pyrophosphatase | |
| NUDT14 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Rabbit | |
| metabolism | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 256281 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:EAWEECGYHLAPSDLRRVATYWSGVGLTGSRQTMFYTEVTDAQRSGPGGGLVEEGELIEVVHLPLEGAQAFADDPDIPKTLGVIFGVSWFLSQ | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title