missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nudel Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00€ - 550.00€
Specifications
| Antigen | Nudel |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30228136
|
Novus Biologicals
NBP3-38185-20ul |
20 μL |
190.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30227124
|
Novus Biologicals
NBP3-38185-100ul |
100 μL |
550.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nudel Polyclonal antibody specifically detects Nudel in Human,Mouse samples. It is validated for ELISA,Western BlotSpecifications
| Nudel | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication, Neuroscience, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 81565 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| DKFZp451M0318, EOPA, MITAP1mitosin-associated protein MITAP1, Mitosin-associated protein 1, nuclear distribution protein nudE-like 1, nudE nuclear distribution gene E homolog (A. nidulans)-like 1, nudE nuclear distribution gene E homolog like 1, nudE nuclear distribution gene E homolog like 1 (A. nidulans), NUDELendooligopeptidase A, NUDE-like protein, Protein Nudel | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-345 of human Nudel (NP_110435.1).,, Sequence:, MDGEDIPDFSSLKEETAYWKELSLKYKQSFQEARDELVEFQEGSRELEAELEAQLVQAEQRNRDLQADNQRLKYEVEALKEKLEHQYAQSYKQVSVLEDDLSQTRAIKEQLHKYVRELEQANDDLERAKRATIVSLEDFEQRLNQAIERNAFLESELDEKESLLVSVQRLKDEARDLRQELAVRERQQEVTRKSAPSSPTLDCEKMDSAVQASLSLPATPVGKGTENTFPSPKAIPNGFGTSPLTPSARISALNIVGDLLRKVGALESKLAACRNFAKDQASRKSYISGNVNCGVLNGNGTKFSRSGHTSFFDKGAVNGFDPAPPPPGLGSSRPSSAPGMLPLSV | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title