missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDCD1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94412-0.1ml
This item is not returnable.
View return policy
Description
NUDCD1 Polyclonal antibody specifically detects NUDCD1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| NUDCD1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Chronic myelogenous leukemia tumor antigen 66, CML66OVA66, FLJ14991, NudC domain containing 1, nudC domain-containing protein 1, Tumor antigen CML66 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 304-583 of human NUDCD1 (NP_116258.2). IQFLPDHINIVLKDHQFLEGKLYSSIDHESSTWIIKESNSLEISLIKKNEGLTWPELVIGDKQGELIRDSAQCAAIAERLMHLTSEELNPNPDKEKPPCNAQELEECDIFFEESSSLCRFDGNTLKTTHVVNLGSNQYLFSVIVDPKEMPCFCLRHDVDALLWQPHSSKQDDMWEHIATFNALGYVQASKRDKKFFACAPNYSYAALCECLRRVFIYRQPAPMSTVLYNRKEGRQVGQVAKQQVASLETNDPILGFQATNERLFVLTTKNLFLIKVNTEN | |
| 0.1 mL | |
| Cancer, Immunology | |
| 84955 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction