Learn More
Macherey-Nagel™ NucleoType™ Mouse PCR
Kit for rapid mouse typing experiments with common samples such as tail clips, ear punch, hair, and blood.
23.10€ - 300.00€
Specifications
Content And Storage | NucleoType HotStart PCR Master Mix (2x), Lysis Buffer M, Proteinase K. Store at 4 °C or -20 °C |
---|---|
Detection Method | Primer-probe |
Format | Kit |
Yield | 100 - 600 ng amplicon per 10 μL PCR |
For Use With (Application) | Genotyping |
Product Code | Brand | No. of Tests | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Tests | Price | Quantity & Availability | |||||
15871907
|
Macherey-Nagel™
743200.25 |
25 Preps |
23.10€
25 reactions |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
15861907
|
Macherey-Nagel™
743200.100 |
100 Preps |
74.60€
100 reactions |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
15881907
|
Macherey-Nagel™
743200.500 |
500 Preps |
300.00€
500 reactions |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
Description
NucleoType™ Mouse PCR
- DNA preparation within 5 minutes
- Loading dye included for subsequent gel electrophoresis
- Designed for tail, ear, blood, and even hair samples
- Simple sample preparation in one single tube combined with a fast HotStart PCR
- Direct PCR for PCR analysis directly from unpurified and undiluted samples
Specifications
NucleoType HotStart PCR Master Mix (2x), Lysis Buffer M, Proteinase K. Store at 4 °C or -20 °C | |
Kit | |
Genotyping | |
Mouse tail clipping (1 mm), mouse ear punch (Dia. 1 mm), mouse blood (1 μL), mouse hairs (approx. 3–30) |
Primer-probe | |
100 - 600 ng amplicon per 10 μL PCR | |
Lysis 2 min; Enzyme inactivation 3 min; PCR cycling 30–90 min | |
Amplicon size: up to 1000 bp |