missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoType™ Mouse PCR
Kit for rapid mouse typing experiments with common samples such as tail clips, ear punch, hair, and blood.
Brand: Macherey-Nagel™ 743200.100
Additional Details : Weight : 0.01600kg
Description
NucleoType™ Mouse PCR
- DNA preparation within 5 minutes
- Loading dye included for subsequent gel electrophoresis
- Designed for tail, ear, blood, and even hair samples
- Simple sample preparation in one single tube combined with a fast HotStart PCR
- Direct PCR for PCR analysis directly from unpurified and undiluted samples
Specifications
NucleoType HotStart PCR Master Mix (2x), Lysis Buffer M, Proteinase K. Store at 4 °C or -20 °C | |
Kit | |
100 - 600 ng amplicon per 10 μL PCR | |
Lysis 2 min; Enzyme inactivation 3 min; PCR cycling 30–90 min | |
Amplicon size: up to 1000 bp |
Primer-probe | |
100 Preps | |
Genotyping | |
Mouse tail clipping (1 mm), mouse ear punch (Dia. 1 mm), mouse blood (1 μL), mouse hairs (approx. 3–30) |