missing translation for 'onlineSavingsMsg'
Learn More

Nucleolin, p Thr76 Antibody (SR1019), Novus Biologicals™

Rabbit Monoclonal Antibody

349.00€ - 449.00€

Specifications

Antigen Nucleolin
Clone SR1019
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18605774
View Documents
Novus Biologicals
NBP3-21607-100ul
100 μg
449.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18655594
View Documents
Novus Biologicals
NBP3-21607-50ul
50 μg
349.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Nucleolin Monoclonal antibody specifically detects Nucleolin in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

Nucleolin
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200
Monoclonal
Purified
RUO
Human
C23, FLJ45706, FLJ59041, nucleolin, Protein C23
A synthesized peptide derived from human phospho-Nucleolin [Thr76] (Uniprot #: P19338)
Primary
Store at -20°C. Avoid freeze/thaw cycles.
SR1019
Western Blot, Immunohistochemistry
Unconjugated
Rabbit
Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Phospho Specific, Stem Cell Markers
PBS (pH 7.4), 150 mM NaCl, 50% Glycerol
4691
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.