missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nuclear Membrane Marker Antibody (AE-5), Novus Biologicals™
Mouse Monoclonal Antibody
252.00€ - 499.00€
Specifications
| Antigen | Nuclear Membrane Marker |
|---|---|
| Clone | AE-5 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunocytochemistry/ Immunofluorescence 1-2 μg/ml |
| Applications | Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18696477
|
Novus Biologicals
NBP3-11307-100ug |
100 μg |
499.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18628077
|
Novus Biologicals
NBP3-11307-20ug |
20 μg |
252.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nuclear Membrane Marker Monoclonal antibody specifically detects Nuclear Membrane Marker in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| Nuclear Membrane Marker | |
| 0.2 mg/mL | |
| Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| Nuclei of myeloid leukemia biopsy cells | |
| Primary | |
| Store at 4°C. |
| AE-5 | |
| Immunocytochemistry/ Immunofluorescence 1-2 μg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title