missing translation for 'onlineSavingsMsg'
Learn More

Nuarimol, TRC

Product Code. 30009967
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009967 25 mg 25mg
30012612 5 mg 5mg
30003709 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009967 Supplier Toronto Research Chemicals Supplier No. TRCN92532025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63284-71-9
Molecular Formula C17 H12 Cl F N2 O
Molecular Weight (g/mol) 314.74
Synonym α-(2-Chlorophenyl)-α-(4-fluorophenyl)-5-pyrimidinemethanol, (±)-Nuarimol, EL 228, EL 228 (fungicide), EL 2289, Nuarimol, Tridal, Trimidal, Trimidal EC, Trimifruit SC
IUPAC Name (2-chlorophenyl)-(4-fluorophenyl)-pyrimidin-5-ylmethanol
SMILES OC(c1ccc(F)cc1)(c2cncnc2)c3ccccc3Cl

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C17H12ClFN2O/c18-16-4-2-1-3-15(16)17(22,13-9-20-11-21-10-13)12-5-7-14(19)8-6-12/h1-11,22H
Formula Weight 314.0622
Chemical Name or Material Nuarimol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.