CAS RN 63284-71-9
CAS RN 63284-71-9
IUPAC Name:
(2-chlorophenyl)(4-fluorophenyl)(pyrimidin-5-yl)methanol
Synonyms:
guantlet nuarimol trimidal trimifruit SC triminol caswell no. 419G murox epa pesticide chemical code 224100 C17H12ClFN2O
Molecular Weight (g/mol):
314.74
Molecular Formula:
C17H12ClFN2O
InChi Key:
SAPGTCDSBGMXCD-UHFFFAOYNA-N
SMILES:
OC(C1=CC=C(F)C=C1)(C1=CN=CN=C1)C1=CC=CC=C1Cl
1
–
1
of
1
results
1
Nuarimol, TRC
CAS: 63284-71-9 Molecular Formula: C17 H12 Cl F N2 O Molecular Weight (g/mol): 314.74 Synonym: α-(2-Chlorophenyl)-α-(4-fluorophenyl)-5-pyrimidinemethanol,(±)-Nuarimol,EL 228,EL 228 (fungicide),EL 2289,Nuarimol,Tridal,Trimidal,Trimidal EC,Trimifruit SC IUPAC Name: (2-chlorophenyl)-(4-fluorophenyl)-pyrimidin-5-ylmethanol SMILES: OC(c1ccc(F)cc1)(c2cncnc2)c3ccccc3Cl