Learn More
nti-NHP CD45 Mouse anti-Rhesus, Cynomolgus, Baboon, BUV805, Clone: D058-1283, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 742055
This item is not returnable.
View return policy
Description
nti-NHP CD45 Monoclonal antibody specifically detects nti-NHP CD45 in Rhesus, Cynomolgus, Baboon samples. It is validated for Flow Cytometry
Specifications
| nti-NHP CD45 | |
| Monoclonal | |
| Brilliant Ultraviolet 805 | |
| Pan Leukocyte, NHP-specific; PTPRC; LCA; L-CA; Leukocyte Common Ag | |
| Rhesus peripheral whole blood | |
| RUO | |
| Rhesus Monkey, Cynomolgus, Baboon | |
| IgG1 κ |
| Flow Cytometry | |
| D058-1283 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV805 under optimal conditions that minimize unconjugated dye and antibody. |