missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NTAL Antibody (NAP-07), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-341H
This item is not returnable.
View return policy
Description
NTAL Monoclonal antibody specifically detects NTAL in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Flow (Intracellular), CyTOF
Specifications
| NTAL | |
| Monoclonal | |
| HRP | |
| LABWilliams-Beuren syndrome chromosomal region 15 protein, Linker for activation of B-cells, linker for activation of T cells family, member 2, linker for activation of T-cells family member 2, Membrane-associated adapter molecule, Non-T-cell activation linker, NTALWilliams-Beuren syndrome chromosomal region 5 protein, WBS15, WBSCR15, WBSCR5HSPC046, Williams-Beuren syndrome chromosome region 5, WSCR5 | |
| Recombinant cytoplasmic domain (amino acids 91-244) of Human NTAL. | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Immunology, Innate Immunity, Signal Transduction, Transcription Factors and Regulators | |
| 7462 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot | |
| NAP-07 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction