missing translation for 'onlineSavingsMsg'
Learn More

NT-proBNP Antibody (16F3), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody

553.00€

Specifications

Antigen NT-proBNP
Clone 16F3
Applications Western Blot
Classification Monoclonal
Conjugate Alexa Fluor 594
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18752052
View Documents
Novus Biologicals
NB200-439AF594
0.2 mL
553.00€
0.20mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NT-proBNP Monoclonal antibody specifically detects NT-proBNP in Human samples. It is validated for Western Blot, ELISA
Specifications

Specifications

NT-proBNP
Western Blot
Alexa Fluor 594
Mouse
Biologically Active Proteins, Cardiovascular Biology, Cytokine Research, Immunology, Neuroscience, Neurotransmission
50mM Sodium Borate
4879
IgG1
Protein A purified
16F3
Monoclonal
Purified
RUO
Human
BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with synthetic NT-proBNP, corresponding to a.a.r. 13-27, conjugated with carrier protein.
Primary
Store at 4°C in the dark.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.