Learn More
NT-proBNP Antibody (11D1cc), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB120-13059DL594
 View more versions of this product
Product Code. 18730733
  • 575.00€ / 0.20mL
Description
NT-proBNP Monoclonal antibody specifically detects NT-proBNP in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NT-proBNP
Monoclonal
DyLight 594
BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with synthetic NT-proBNP, corresponding to a.a.r. 28-45, conjugated with carrier protein.
0.2 mL
Biologically Active Proteins, Cardiovascular Biology, Cytokine Research, Immunology, Neuroscience, Neurotransmission
4879
Store at 4°C in the dark.
IgG1
Western Blot
11D1cc
50mM Sodium Borate
Mouse
Protein A purified