missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSUN3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00€
Specifications
| Antigen | NSUN3 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
NSUN3 Polyclonal specifically detects NSUN3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| NSUN3 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 63899 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:KQYSKELGDAWNTVREILTSPSCWQYAVLLNRFNYPFELEKDLHLKGYHTLSQGSLPNYPKSVKCYLSRTPGRIPSERHQIGNLKKYYLLNAASLL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| EC 2.1.1, EC 2.1.1.-, FLJ22109, FLJ22609, MST077, NOL1/NOP2/Sun domain family 3, NOL1/NOP2/Sun domain family member 3, NOL1/NOP2/Sun domain family, member 3, NOP2/Sun domain family, member 3, putative methyltransferase NSUN3 | |
| NSUN3 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title