missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nse2 Antibody (OTI6A6), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73070H
This item is not returnable.
View return policy
Description
Nse2 Monoclonal specifically detects Nse2 in Human samples. It is validated for Western Blot.
Specifications
| Nse2 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| NSMCE2 | |
| Full length recombinant protein of human Nse2 (NP_775956) produced in E.coli. | |
| 0.1 mL | |
| 286053.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI6A6 | |
| Western Blot | |
| C8orf36, E3 SUMO-protein ligase NSE2, EC 6.3.2, EC 6.3.2.-, FLJ32440, hMMS21, methyl methanesulfonate sensitivity gene 21, MMS21 homolog, MMS21chromosome 8 open reading frame 36, Non-SMC element 2 homolog, non-SMC element 2 homolog (MMS21, S. cerevisiae), non-SMC element 2, MMS21 homolog (S. cerevisiae), Non-structural maintenance of chromosomes element 2 homolog, NSE2 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction