missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nrf2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-32822
This item is not returnable.
View return policy
Description
Nrf2 Polyclonal antibody specifically detects Nrf2 in Human, Mouse, Rat, Alligator, Avian, Plant, Zebrafish samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Knockdown.
Specifications
| Nrf2 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH7), 20% Glycerol with 0.025% Proclin 300 | |
| HEBP1, NF-E2-related factor 2, NFE2-related factor 2, NRF2 erythroid derived 2, like 2, Nrf2 ihc, Nrf2 immunofluorescence, Nrf2 immunohistochemistry, Nrf2 polyclonal, Nrf2 rat, Nrf2 western blot, nuclear factor (erythroid-derived 2)-like 2, nuclear factor erythroid 2-related factor 2, nuclear factor erythroid-derived 2-like 2 | |
| Rabbit | |
| 68 kDa | |
| 0.1 mL | |
| Transcription Factors and Regulators | |
| 4780 | |
| Human, Mouse, Rat, Alligator, Avian, Bovine, C. elegans, Plant | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Simple Western reported by internal validation, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Knockdown Validated | |
| Q16236 | |
| NFE2L2 | |
| Recombinant protein encompassing a sequence within the center region of human NRF2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Predicted cross-reactivity based on sequence identity: Cow (85%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction