Learn More
NR1D2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13906-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the nuclear hormone receptor family, specifically the NR1 subfamily of receptors. The encoded protein functions as a transcriptional repressor and may play a role in circadian rhythms and carbohydrate and lipid metabolism. Alternatively spliced transcript variants have been described.Specifications
| NR1D2 | |
| Polyclonal | |
| Unconjugated | |
| NR1D2 | |
| BD73, EAR 1R, NR1D2, Rev erb beta, Reverb Beta, RVR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 259241, 353187, 9975 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q14995, Q60674, Q63504 | |
| Nr1d2 | |
| NR1D2 Fusion Protein Ag4881 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |