missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPTN Monoclonal Antibody (OTI2D4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504330S
This item is not returnable.
View return policy
Description
NPTN Monoclonal antibody specifically detects NPTN in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| NPTN | |
| Monoclonal | |
| 0.69 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P49069 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 819 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2D4 | |
| Unconjugated | |
| CAMLG | |
| AW554172; glycoprotein 55; glycoprotein 55/65; glycoprotein 65; GP55; gp55/65; GP65; Neuroplastin; np55; np65; NPTN; Sdfr1; Sdr1; SDR-1; stromal cell derived factor receptor 1; stromal cell-derived receptor 1 | |
| Full length human recombinant protein of human CAMLG produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction