missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Npas4 Antibody (S408-79), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
188.00€ - 614.00€
Specifications
| Antigen | Npas4 |
|---|---|
| Clone | S408-79 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Frozen |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18075234
|
Novus Biologicals
NBP2-59332 |
100 μg |
614.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18687158
|
Novus Biologicals
NBP2-59332-25ug |
25 μg |
188.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Npas4 Monoclonal specifically detects Npas4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
| Npas4 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| BHLHE79, bHLHe79neuronal PAS domain-containing protein 4, Class E basic helix-loop-helix protein 79, Le-PAS, neuronal PAS domain protein 4, Neuronal PAS4, NXFHLH-PAS transcription factor NXF, PASD10PAS domain-containing protein 10 | |
| NPAS4 | |
| IgG1 | |
| Protein G purified |
| S408-79 | |
| Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Frozen | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| 266743 | |
| Fusion protein amino acids 597-802 (C-terminus) of rat Npas4. | |
| Primary | |
| Detects 90kDa. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title