missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Noxa Antibody (69N15C9), Alexa Fluor™ 700, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-25266AF700
This item is not returnable.
View return policy
Description
Noxa Monoclonal specifically detects Noxa in Mouse samples. It is validated for Western Blot.
Specifications
| Noxa | |
| Monoclonal | |
| Alexa Fluor 700 | |
| adult T cell leukemia-derived PMA-responsive, APR, Immediate-early-response protein APR, NOXAPMA-induced protein 1, phorbol-12-myristate-13-acetate-induced protein 1, PMA-Induced Protein 1, Protein Noxa | |
| Rat | |
| 11.6 kDa | |
| 0.1 mL | |
| Apoptosis, Cancer, Cellular Markers, Membrane Trafficking and Chaperones, Plasma Membrane Markers, Stem Cell Markers, Tumor Suppressors | |
| 5366 | |
| Mouse | |
| Purified |
| Western Blot | |
| 69N15C9 | |
| Western Blot | |
| PMAIP1 | |
| This Noxa antibody was developed by immunizing mice with a fusion protein containing human Noxa. | |
| Protein G purified | |
| RUO | |
| Primary | |
| This product reacts with Mouse. | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction