missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nox4 Antibody (NOX4/1245), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54338IR
This item is not returnable.
View return policy
Description
Nox4 Monoclonal antibody specifically detects Nox4 in Human samples. It is validated for Western Blot, ELISA, CyTOF
Specifications
| Nox4 | |
| Monoclonal | |
| DyLight 755 | |
| EC 1.6.3, Kidney oxidase-1, Kidney superoxide-producing NADPH oxidase, KOX, KOX-1Renal NAD(P)H-oxidase, NADPH oxidase 4, RENOXEC 1.6.3.- | |
| Recombinant human Nox4 protein (Uniprot: Q9NPH5) | |
| 100 μg | |
| Cancer, Cell Biology, Diabetes Research, Hypoxia, Lipid and Metabolism, Signal Transduction | |
| 50507 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, ELISA, CyTOF | |
| NOX4/1245 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction