missing translation for 'onlineSavingsMsg'
Learn More

NOX3 Antibody - BSA Free, Novus Biologicals™

Product Code. 18652768 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.03mg
0.10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18652768 0.025 mg 0.03mg
18031884 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18652768 Supplier Novus Biologicals Supplier No. NBP2412920.025mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NOX3 Polyclonal specifically detects NOX3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOX3
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Alias EC 1.6.3, GP91-3EC 1.6.3.-, gp91phox homolog 3, Mitogenic oxidase 2, MOX2, MOX-2, NADPH oxidase 3, NADPH oxidase catalytic subunit-like 3
Gene Symbols NOX3
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid peptide near the amino terminus of human NOX3. The immunogen is located within amino acids 110 - 160 of NOX3. Amino Acid Squence: NLERYHWSQSEEAQG
Molecular Weight of Antigen 72 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50508
Test Specificity NOX3 is predicted to not cross-react with other NOX proteins.
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.