missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOX1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93835-0.1ml
This item is not returnable.
View return policy
Description
NOX1 Polyclonal antibody specifically detects NOX1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| NOX1 | |
| Polyclonal | |
| Western Blot 1:200 - 1:500, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| EC 1.6.3, GP91-2, Mitogenic oxidase 1, MOX-1, MOX1NADH/NADPH mitogenic oxidase subunit P65-MOX, NADPH oxidase 1, NADPH oxidase homolog-1, NOH1mitogenic oxidase (pyridine nucleotide-dependent superoxide-generating), NOH-1NADPH oxidase 1 variant NOH-1L, NOX-1 | |
| A synthetic peptide corresponding to a sequence within amino acids 418-564 of human NOX1 (NP_008983.2). WYKFQCADHNLKTKKIYFYWICRETGAFSWFNNLLTSLEQEMEELGKVGFLNYRLFLTGWDSNIVGHAALNFDKATDIVTGLKQKTSFGRPMWDNEFSTIATSHPKSVVGVFLCGPRTLAKSLRKCCHRYSSLDPRKVQFYFNKENF | |
| 0.1 mL | |
| Cancer, Cell Biology, Direct Reversal of DNA Damage, Endocrinology, Immunology, Signal Transduction | |
| 27035 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction