Learn More
Tris-EDTA Buffer Solutions, Novus Biologicals™
Shop All Bio Techne Products
Click to view available options
pH:
8
9
Unit Size:
500mL
Description
- 10X EDTA buffer for Immunohistochemistry
- Dilute one part buffer with nine parts distilled water
- Unit size: 500mL
Recommended Storage
Store at room temperature
Specifications
Specifications
| For Use With (Application) | Retrieval of antigens masked by fixation in cross linking fixatives such as formalin |
| pH | 9 |
| Quantity | 500 mL |