missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ UROD Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.1mg
0.50mg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-367 of Human UROD The Recombinant Human UROD Protein is derived from E. coli. The Recombinant Human UROD Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 0.5mg/mL |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | In 20mM Tris-HCl buffer (pH 8.0) containing 20% glycerol 1mM DTT, 0.1M NaCl, and 1mM EDTA. |
| Gene ID (Entrez) | 7389 |
| Molecular Weight (g/mol) | 43.0kDa |
| Purification Method | Protein |
| Quantity | 0.1 mg |
| Source | Human |
| Immunogen | MGSSHHHHHH SSGLVPRGSH MEANGLGPQG FPELKNDTFL RAAWGEETDY TPVWCMRQAG RYLPEFRETR AAQDFFSTCR SPEACCELTL QPLRRFPLDA AIIFSDILVV PQALGMEVTM VPGKGPSFPE PLREEQDLER LRDPEVVASE LGYVFQAITL TRQRLAGRVP LIGFAGAPWT LMTYMVEGGG SSTMAQAKRW LYQRPQASHQ LLRILTDALV PYLVGQVVAG AQALQLFESH AGHLGPQLFN KFALPYIRDV AKQVKARLRE AGLAPVPMII FAKDGHFALE ELAQAGYEVV GLDWTVAPKK ARECVGKTVT LQVNLDPCAL YASEEEIGQL VKQMLDDFGP HRYIANLGHG LYPDMDPEHV GAFVDAVHKH SRLLRQN |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction