missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human NSMCE1 His Protein

Product Code. 18106431 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18106431 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18106431 Supplier Novus Biologicals™ Supplier No. NBP223292

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-266 of Human NSMCE1 The Recombinant Human NSMCE1 Protein is derived from E. coli. The Recombinant Human NSMCE1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_659547
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.15 M NaCl, 20% glycerol, 1mM DTT
Gene ID (Entrez) 197370
Molecular Weight (g/mol) 33kDa
Name NSMCE1 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen NSMCE1, 1-266aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMQGSTRR MGVMTDVHRR FLQLLMTHGV LEEWDVKRLQ THCYKVHDRN ATVDKLEDFI NNINSVLESL YIEIKRGVTE DDGRPIYALV NLATTSISKM ATDFAENELD LFRKALELII DSETGFASST NILNLVDQLK GKKMRKKEAE QVLQKFVQNK WLIEKEGEFT LHGRAILEME QYIRETYPDA VKICNICHSL LIQGQSCETC GIRMHLPCVA KYFQSNAEPR CPHCNDYWPH EIPKVFDPEK ERESGVLKSN KKSLRSRQH
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.