missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Myosin Light Chain 2 His Protein
Shop All Bio Techne Products
Click to view available options
:
0.1mg; Unlabeled
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids1-166 of Human Myosin Light Chain 2 The Recombinant Human Myosin Light Chain 2 Protein is derived from E. coli. The Recombinant Human Myosin Light Chain 2 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | NP_000423 |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH 8.0), 40% glycerol, 5mM CaCl2 |
| Gene ID (Entrez) | 4633 |
| Molecular Weight (g/mol) | 20.9kDa |
| Name | Myosin Light Chain 2 Protein |
| Purification Method | Protein |
| Immunogen | MGSSHHHHHHSSGLVPRGSHMAPKKAKKRAGGANSNVFSMFEQTQIQEFKEAFTIMDQNRDGFIDKNDLRDTFAALGRVNVKNEEIDEMIKEAPGPINFTVFLTMFGEKLKGADPEETILNAFKVFDPEGKGVLKADYVREMLTTQAERFSKEEVDQMFAAFPPDVTGNLDYKNLVHIITHGEEKD |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Endotoxin Concentration | <1 EU per 1ug of protein (determined by LAL method) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction