Learn More
Novus Biologicals™ Recombinant Human HLA C His Protein
Product Code. 18796913 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Product Code. 18796913
Brand: Novus Biologicals™ NBP223210
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 25-308 of Human HLA C The Recombinant Human HLA C Protein is derived from E. coli. The Recombinant Human HLA C Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 0.1 mg