missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human CRABP2 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Description
An un-tagged recombinant protein corresponding to the amino acids1-138 of Human E.coli CRABP2 The Recombinant Human CRABP2 Protein is derived from E. coli. The Recombinant Human CRABP2 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | NP_001869 |
| Concentration | 1 mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | 20 mM Tris-HCl buffer (pH 8.0), 20% glycerol |
| Gene ID (Entrez) | 1382 |
| Molecular Weight (g/mol) | M.W. (theoretical): 15.6 kDa |
| Name | CRABP2 Protein |
| Purification Method | Protein |
| Quantity | 0.1 mg |
| Immunogen | MPNFSGNWKIIRSENFEELLKVLGVNVMLRKIAVAAASKPAVEIKQEGDTFYIKTSTTVRTTEINFKVGEEFEEQTVDGRPCKSLVKWESENKMVCEQKLLKGEGPKTSWTRELTNDGELILTMTADDVVCTRVYVRE |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction