Learn More
Novus Biologicals™ Recombinant Human alpha-Smooth Muscle Actin His Protein
Product Code. 18760414 Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
Product Code. 18760414
Brand: Novus Biologicals™ NBP222670
Highly purified. Generating reliable and reproducible results.
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 3-377 of Human alpha-Smooth Muscle Actin The Recombinant Human alpha-Smooth Muscle Actin Protein is derived from E. coli. The Recombinant Human alpha-Smooth Muscle Actin Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 0.05 mg