Learn More
NOS2 Monoclonal Antibody (OTI3H7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813521
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
NOS2 Monoclonal antibody specifically detects NOS2 in Human samples. It is validated for Western BlotSpecifications
| NOS2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P35228 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 4843 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI3H7 | |
| Unconjugated | |
| Nos2 | |
| Hepatocyte NOS; hepatocytes; HEP-NOS; inducible nitric oxide synthase; inducible NO synthase; Inducible NOS; iNos; i-NOS; Inosl; MAC-NOS; macrophage NOS; nitric oxide synthase 2; nitric oxide synthase 2, inducible; nitric oxide synthase 2, inducible, macrophage; nitric oxide synthase 2A (inducible, hepatocytes); nitric oxide synthase, inducible; nitric oxide synthase, macrophage; nitric oxide synthase-inducible; NOS; NOS type II; NOS, type II; Nos2; Nos-2; Nos2a; NOS-II; OTTMUSP00000000202; peptidyl-cysteine S-nitrosylase NOS2 | |
| Human recombinant protein fragment corresponding to amino acids 539-970 of human Nos2 (NP_000616) produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.