missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Norpheniramine Maleate, Mikromol™
Description
Norpheniramine Maleate
Specifications
Specifications
| Chemical Name or Material | Norpheniramine Maleate |
| CAS | 19428-44-5 |
| Analyte or Component Names | Norpheniramine Maleate |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C15 H18 N2 . C4 H4 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Norpheniramine Maleate, N-Methyl-gamma-phenyl-2-pyridinepropanamine Maleate, 2-[alpha-[2-(Methylamino)ethyl]benzyl]-pyridine Maleate, Monodesmethylpheniramine Maleate, N-Demonomethylpheniramine Maleate |
| InChI Formula | InChI=1S/C15H18N2.C4H4O4/c1-16-12-10-14(13-7-3-2-4-8-13)15-9-5-6-11-17-15;5-3(6)1-2-4(7)8/h2-9,11,14,16H,10,12H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1- |
| SMILES | CNCCC(c1ccccc1)c2ccccn2.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;N-methyl-3-phenyl-3-pyridin-2-ylpropan-1-amine |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?