missing translation for 'onlineSavingsMsg'
Learn More

Nordiphenhydramine Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal

Product Code. 30458602
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30458602

Brand: LoGiCal LGCAMP0490.0301

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Nordiphenhydramine Hydrochloride 1.0 mg/ml in Methanol (as free base)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nordiphenhydramine Hydrochloride
CAS 53499-40-4
Analyte or Component Names Nordiphenhydramine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C16 H19 N O . Cl H
Certifications/Compliance ISO 17034
Synonym 2-(Diphenylmethoxy)-N-methylethanamine Hydrochloride (Nordiphenhydramine Hydrochloride), Dimenhydrinate Imp. F (EP), Diphenhydramine Hydrochloride Imp. A (EP), Diphenhydramine Imp. A (EP), Diphenhydramine USP Related Compound A, Diphenhydramine USP RC A, 2-(Diphenylmethoxy)-N-methylethanamine Hydrochloride, Nordiphenhydramine Hydrochloride, Dimenhydrinate Imp. F (EP) as Hydrochloride, Diphenhydramine Imp. A Hydrochloride, Diphenhydramine Hydrochloride Imp. A (EP) as Hydrochloride, Diphenhydramine Related Compound A as Hydrochloride, Dimenhydrinate Impurity F as Hydrochloride, Diphenhydramine Hydrochloride Impurity A as Hydrochloride, Diphenhydramine Impurity A as Hydrochloride
InChI Formula InChI=1 S/C16H19NO.ClH/c1-17-12-13-18-16(14-8-4-2-5-9-14)15-10-6-3-7-11-15;/h2-11,16-17 H,12-13H2,1H3;1 H
SMILES Cl.CNCCOC(c1ccccc1)c2ccccc2
IUPAC Name 2-benzhydryloxy-N-methylethanamine;hydrochloride
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 277.79
Quantity 1 mL
Formula Weight 277.1233 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.