Learn More
Nonenylsuccinic anhydride, 96%
Product Code. 11448237
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11448237 250 g 250g
11438237 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11448237 Supplier Thermo Scientific Alfa Aesar Supplier No. A17607.30
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
CAS: 28928-97-4 | C13H20O3
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 28928-97-4
Molecular Formula C13H20O3
MDL Number MFCD00014552
Specifications
Density 1.032
Boiling Point 126°C to 130°C (0.15 mmHg)
Flash Point 146°C (295°F)
Quantity 250 g
Sensitivity Moisture sensitive
Formula Weight 224.3
Percent Purity 96%
Chemical Name or Material Nonenylsuccinic anhydride