accessibility menu, dialog, popup

UserName

H3COOO(E)

CAS RN 28928-97-4

IUPAC Name: 3-[(1E)-non-1-en-1-yl]oxolane-2,5-dione
Synonyms: 3-[(1E)-non-1-en-1-yl]oxolane-2,5-dione
Molecular Weight (g/mol): 224.3
Molecular Formula: C13H20O3
InChi Key: FGQUIQAGZLBOGL-CMDGGOBGNA-N
SMILES: CCCCCCC\C=C\C1CC(=O)OC1=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)