missing translation for 'onlineSavingsMsg'
Learn More

NOMO2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Specifications

Antigen NOMO2
Concentration 0.23 mg/mL
Applications Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16852914
View Documents
Proteintech
14328-1-AP-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16842914
View Documents
Proteintech
14328-1-AP-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NOMO2
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence
Unconjugated
Rabbit
Human
Q5JPE7
283820
NOMO2 Fusion Protein Ag5595
Primary
-20°C
NOMO2
0.23 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
NODAL modulator 2, Nomo, NOMO2, PM5, pM5 protein 2
NOMO2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.