missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOMO2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94729-0.1ml
This item is not returnable.
View return policy
Description
NOMO2 Polyclonal antibody specifically detects NOMO2 in Human samples. It is validated for Western Blot
Specifications
| NOMO2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| NODAL modulator 2, NOMO, PM5, pM5 protein 2, pM5 protein, centromeric copy | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 873-1222 of human NOMO2 (NP_001004060.1). YALAGVSFEIKAEDDQPLPGVLLSLSGGLFRSNLLTQDNGILTFSNLSPGQYYFKPMMKEFRFEPSSQMIEVQEGQNLKITITGYRTAYSCYGTVSSLNGEPEQGVAMEAVGQNDCSIYGEDTVTDEEGKFRLRGLLPGCVYHVQLKAEGNDHIERALPHHRVIEVGNNDIDDVNIIVFRQINQFDLSGNVITSSEYLPTLWVKLYKSENLDNPIQTVSLGQSLFFHFPPLLRDGENYVVLLDSTLPRSQYDYILPQVSFTAVGYHKHITLIFNPTRKLPEQDIAQGSYIALPLTLLVLLAGYNHDKLIPLLLQLTSRLQGVGALGQAASDNSGPEDAKRQAKKQKTRRT | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 283820 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction