missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOL6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00€ - 550.00€
Specifications
| Antigen | NOL6 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30232624
|
Novus Biologicals
NBP3-35433-20ul |
20 μL |
190.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30231675
|
Novus Biologicals
NBP3-35433-100ul |
100 μL |
550.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NOL6 Polyclonal antibody specifically detects NOL6 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| NOL6 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| bA311H10.1, FLJ21959, MGC14896, MGC14921, MGC20838, NRAP, nucleolar protein 6, nucleolar protein family 6 (RNA-associated), Nucleolar RNA-associated protein, UTP22 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1032-1146 of human NOL6 (NP_075068.2).,, Sequence:, SFCRGLLSQPGPSSLMPVLGYDPPQLYLTQLREAFGDLALFFYDQHGGEVIGVLWKPTSFQPQPFKASSTKGRMVMSRGGELVMVPNVEAILEDFAVLGEGLVQTVEARSERWTV | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 65083 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title