missing translation for 'onlineSavingsMsg'
Learn More

NOC4L Mouse anti-Human, Clone: PCRP-NOC4L-1E3, Novus Biologicals™

Mouse Monoclonal Antibody

484.00€

Specifications

Antigen NOC4L
Clone PCRP-NOC4L-1E3
Concentration 0.2 mg/mL
Dilution Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array
Applications Flow Cytometry, Immunofluorescence, Peptide Array
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18635177
View Documents
Novus Biologicals
NBP3-07362-100ug
100 μg
484.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NOC4L Monoclonal specifically detects NOC4L in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.
Specifications

Specifications

NOC4L
0.2 mg/mL
Flow Cytometry, Immunofluorescence, Peptide Array
Unconjugated
Mouse
Human
MGC3162, NET49, NOC4, NOC4 protein homolog, NOC4-like protein, nucleolar complex associated 4 homolog (S. cerevisiae), nucleolar complex protein 4 homolog, Nucleolar complex-associated protein 4-like protein
Recombinant full-length human NOC4L protein
Primary
Store at 4°C.
PCRP-NOC4L-1E3
Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
79050
IgG1
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.