missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMNAT-2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94693-0.02ml
This item is not returnable.
View return policy
Description
NMNAT-2 Polyclonal antibody specifically detects NMNAT-2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NMNAT-2 | |
| Polyclonal | |
| Western Blot 1:100 - 1:500 | |
| C1orf15, chromosome 1 open reading frame 15, EC 2.7.7.1, EC 2.7.7.18, KIAA0479MGC2756, NaMN adenylyltransferase 2, nicotinamide mononucleotide adenylyltransferase 2, nicotinamide nucleotide adenylyltransferase 2, Nicotinate-nucleotide adenylyltransferase 2, NMN adenylyltransferase 2, PNAT2, pyridine nucleotide adenylyltransferase 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-302 of human NMNAT2 (NP_733820.1). MEIQELEEIQACQGLWEVFVTLSERARDYLHKTGRFIVIGGIVSPVHDSYGKQGLVSSRHRLIMCQLAVQNSDWIRVDPWECYQDTWQTTCSVLEHHRDLMKRVTGCILSNVNTPSMTPVIGQPQNETPQPIYQNSNVATKPTAAKILGKVGESLSRICCVRPPVERFTFVDENANLGTVMRYEEIELRILLLCGSDLLESFCIPGLWNEADMEVIVGDFGIVVVPRDAADTDRIMNHSSILRKYKNNIMVVKDDINHPMSVVSSTKSRLALQHGDGHVVDYLSQPVIDYILKSQLYINASG | |
| 0.02 mL | |
| Endocrinology, Signal Transduction | |
| 23057 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction