missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NME3 (Human) Recombinant Protein
Human NME3 full-length ORF ( AAH00250, 1 a.a. - 169 a.a.) recombinant protein with GST-tag at N-terminal.
335.00€ - 508.00€
Specifications
Accession Number | AAH00250 |
---|---|
Gene ID (Entrez) | 4832 |
Name | non-metastatic cells 3, protein expressed in |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16148411
|
Abnova™
H00004832-P01.10UG |
10 μg |
335.00€
10µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
16158411
|
Abnova™
H00004832-P01.25UG |
25 μg |
508.00€
25µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
- Sequence: MICLVLTIFANLFPAACTGAHERTFLAVKPDGVQRRLVGEIVRRFERKGFKLVALKLVQASEELLREHYAELRERPFYGRLVKYMASGPVVAMVWQGLDVVRTSRALIGATNPADAPPGTIRGDFCIEVGKNLIHGSDSVESARREIALWFRADELLCWEDSAGHWLYE
Specifications
AAH00250 | |
non-metastatic cells 3, protein expressed in | |
125% SDS-PAGE Stained with Coomassie Blue | |
DR-nm23, KIAA0516, NDPK-C, NDPKC, NM23-H3, c371H6.2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
4832 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NME3 | |
GST |