missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NME3 (Human) Recombinant Protein
Human NME3 full-length ORF ( AAH00250, 1 a.a. - 169 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004832-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MICLVLTIFANLFPAACTGAHERTFLAVKPDGVQRRLVGEIVRRFERKGFKLVALKLVQASEELLREHYAELRERPFYGRLVKYMASGPVVAMVWQGLDVVRTSRALIGATNPADAPPGTIRGDFCIEVGKNLIHGSDSVESARREIALWFRADELLCWEDSAGHWLYE
Specifications
AAH00250 | |
non-metastatic cells 3, protein expressed in | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NME3 | |
GST |
4832 | |
Wheat germ expression system | |
10 μg | |
DR-nm23, KIAA0516, NDPK-C, NDPKC, NM23-H3, c371H6.2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |