missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMDAR2C, p Ser1096 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-29516
This item is not returnable.
View return policy
Description
NMDAR2C Polyclonal specifically detects NMDAR2C in Mouse, Rat samples. It is validated for Western Blot.
Specifications
| NMDA R, NR2C Subunit (p Ser1096) | |
| Polyclonal | |
| Western Blot 1:1000 | |
| Q00961 | |
| GRIN2C | |
| Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser1096 conjugated to KLH. Accession # Q00961 | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Specific for the ∼140k NR2C subunit of the NMDA receptor phosphorylated at Ser1096. Immunolabeling is blocked by preadsorption of antibody with the phospho-peptide that was used to generate the antibody but not by the corresponding dephospho-peptide. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
| glutamate [NMDA] receptor subunit epsilon-3, glutamate receptor, ionotropic, N-methyl D-aspartate 2C, NMDAR2CNR2C, N-methyl D-aspartate receptor subtype 2C, N-methyl-D-aspartate receptor subunit 2C | |
| Rabbit | |
| 140 kDa | |
| 0.1 mL | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 2905 | |
| Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction