missing translation for 'onlineSavingsMsg'
Learn More

NMDAR1 Antibody (R1JHL), R&D Systems™

Product Code. 18719913 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.015 mg
Unit Size:
15µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18719913 0.015 mg 15µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18719913 Supplier Novus Biologicals Supplier No. NB300118
  • ONLINE EXCLUSIVE PRICE
    613.00€
    579.60€ / 15µg
    Save 33.40€
    5% Off
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody has been used in 13 publications

NMDAR1 Monoclonal specifically detects NMDAR1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMDAR1
Applications Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone R1JHL
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 3ug/200ug lysate, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1 ug per ml
Gene Accession No. P35439
Gene Alias glutamate [NMDA] receptor subunit zeta-1, glutamate receptor, ionotropic, N-methyl D-aspartate 1, NMDA1, NMDAR1glutamate [NMDA] receptor subunit zeta 1, NMD-R1, N-methyl-D-aspartate receptor channel, subunit zeta-1, N-methyl-D-aspartate receptor subunit NR1, NR1
Gene Symbols GRIN1
Host Species Mouse
Immunogen Fusion protein containing amino acids 1-564 of the NMDAR1 subunit. Accession # P35439
Molecular Weight of Antigen 120 kDa
Purification Method Tissue culture supernatant
Quantity 0.015 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2902
Test Specificity Specific for the ∼120k NR1 subunit of the NMDA receptor.
Reconstitution Reconstitute with 50 ul PBS to desired concentration.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Supernatant
Isotype IgG
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.