missing translation for 'onlineSavingsMsg'
Learn More

NMDAR1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 2 publications

Brand:  Novus Biologicals NB100-41105

Product Code. 18056554

  • 543.00€ / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NMDAR1 Polyclonal specifically detects NMDAR1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NMDAR1
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
glutamate [NMDA] receptor subunit zeta-1, glutamate receptor, ionotropic, N-methyl D-aspartate 1, NMDA1, NMDAR1glutamate [NMDA] receptor subunit zeta 1, NMD-R1, N-methyl-D-aspartate receptor channel, subunit zeta-1, N-methyl-D-aspartate receptor subunit NR1, NR1
Goat
105 kDa
0.1 mg
Neuronal Cell Markers, Neuroscience, Neurotransmission
2902
Human, Rat
IgG
Immunocytochemistry, Immunofluorescence, ELISA
0.5 mg/mL
Immunocytochemistry/Immunofluorescence 10 ug/mL
Q05586
GRIN1
Peptide with sequence C-TQERVNNSNKKE corresponding to internal region according to NP_067544.1, NP_015566.1.
Affinity Purified
RUO
Primary
This antibody is expected to recognize all three reported isoforms (NP_000823.4; NP_067544.1; NP_015566.1).
Store at -20C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only