missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMDAR1 Antibody (24F5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26543-50ul
This item is not returnable.
View return policy
Description
NMDAR1 Monoclonal antibody specifically detects NMDAR1 in Human samples. It is validated for Flow Cytometry, ELISA
Specifications
| NMDAR1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA | |
| 24F5 | |
| Flow Cytometry 1:50-1:200, ELISA | |
| glutamate [NMDA] receptor subunit zeta-1, glutamate receptor, ionotropic, N-methyl D-aspartate 1, NMDA1, NMDAR1glutamate [NMDA] receptor subunit zeta 1, NMD-R1, N-methyl-D-aspartate receptor channel, subunit zeta-1, N-methyl-D-aspartate receptor subunit NR1, NR1 | |
| A synthesized peptide derived from Human NMDAR1 [UniProt Q05586] | |
| 50 μL | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific | |
| 2902 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction