missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NM23-H2/NME2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19552
This item is not returnable.
View return policy
Description
NM23-H2/NME2 Polyclonal specifically detects NM23-H2/NME2 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| NM23-H2/NME2 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 40% Glycerol with 0.01% Thimerosal | |
| NME2 | |
| Recombinant protein encompassing a sequence within the center region of human nm23-H2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse. Species reactivity based on sequence identity: Rabbit (100%), Rat (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| EC 2.7.13.3, Histidine protein kinase NDKB, MGC2212, NDK B, NDKB, NDPKB, NDPK-B, nm23-H2, non-metastatic cells 2, protein (NM23B) expressed in, nucleoside diphosphate kinase B, protein (NM23) expressed in | |
| Rabbit | |
| 17 kDa | |
| 0.1 mL | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 4831 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction