missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLRX1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
588.00€
Specifications
| Antigen | NLRX1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 500 SE |
| Form | Purified |
Description
NLRX1 Polyclonal antibody specifically detects NLRX1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| NLRX1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Caterpiller protein 11.3, CLR11.3DLNB26, MGC131937, MGC21025, NLR family member X1, NLR family, X1, NOD26, NOD5, NOD9FLJ21478, NOD-like receptor X1, Nucleotide-binding oligomerization domain protein 26, Nucleotide-binding oligomerization domain protein 5, Nucleotide-binding oligomerization domain protein 9, nucleotide-binding oligomerization domain, leucine rich repeat containing X1 | |
| Antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of human NOD5. The immunogen is located within amino acids 70 - 120 of NOD5. Amino Acid Squence: FSRLPREERQFGPT | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot | |
| mFluor Violet 500 SE | |
| Rabbit | |
| Immunology, Innate Immunity | |
| 50mM Sodium Borate | |
| 79671 | |
| IgG | |
| Peptide affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title